Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Ultra Precise Manipulator Arm
SDP

Catalog No. 10000399 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Digital manipulator arm, Left
Digital manipulator arm, Right
Manipulator arm, Left
Manipulator arm, Right
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-399 Manipulator arm, Right
10-000-397 Manipulator arm, Left
10-000-421 Digital manipulator arm, Left
10-000-423 Digital manipulator arm, Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-399 Supplier Stoelting™ Supplier No. 51606U
Add to Cart
Add to Cart
Request a Quote

Ideal for use in mice and rats where high-accuracy positioning is required

  • Up to 80 mm of travel is available in all directions
  • Up to 90° angle adjustment in either the anterior-posterior or medial-lateral planes
  • Compatible with all Stoelting Stereotaxic Series including New Standard, Lab Standard, Just for Mice and Just for Mouse Dual, Digital and Motorized Ultra-Precise Manipulators are also available.
  • 10 micron resolution
  • Target small brain regions with confidence

Specifications

For Use With (Application) Neuroscience
Size 3 axis
Animal Mouse, Rat
Model Manipulator arm, Right
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.