Learn More
SCP Science Nitric acid, PPP, 67 - 70%, 500ml, 7697-37-2
Supplier: SCP Science 250036131
PlasmaPURE Plus acids and reagents are manufactured with trace metal impurity levels less than 10 ppt (0.01ppb) for ultra low level analysis below 1ppb. Supplied in preleached Teflon bottles for optimum quality and packaged in Class 10 clean room conditions. Complete with a Certificate of Analysis with lot number, expiry date and maximum specification for over 60 analytes. Hazardous: UN Code 2031, Nitric acid ** Cargo Only **, Packing Group II, Class 8(5.1)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.