Learn More
Promega Ultra Pure UDP-GalNAc, 100mM, 250microl
Catalog No. PRV7082 Shop All Promega Corporation Products
Click to view available options
Quantity:
250 μL
UDP-sugar substrates are used with the UDP-Glo Glycosyltransferase Assay to detect the activity of glycosyltransferases that use UDP-sugars as donor substrates and release UDP as a product.
  • 100mM UDP−Sugar Substrates for use with the UDP−Glo™ Glycosyltransferase Assay
Specifications
Content And Storage <-65°C
Quantity 250 μL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.