Learn More
Promega Ultra Pure UDP-Glucose, 100mM, 250microl
Catalog No. PRV7092 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
250 μL
Catalog No. Quantity
PRV7092 250 μL
1 options
Catalog No. PRV7092 Supplier Promega Supplier No. V7092
UDP-sugar substrates are used with the UDP-Glo Glycosyltransferase Assay to detect the activity of glycosyltransferases that use UDP-sugars as donor substrates and release UDP as a product.
  • 100mM UDP−Sugar Substrates for use with the UDP−Glo™ Glycosyltransferase Assay
Specifications
Content And Storage <-65°C
Quantity 250 μL