Learn More
Promega Ultra Pure UDP-Glucose, 100mM, 50microl
Catalog No. PRV7091 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PRV7091 50 μL
1 options
Catalog No. PRV7091 Supplier Promega Supplier No. V7091
UDP-sugar substrates are used with the UDP-Glo Glycosyltransferase Assay to detect the activity of glycosyltransferases that use UDP-sugars as donor substrates and release UDP as a product.
  • 100mM UDP−Sugar Substrates for use with the UDP−Glo™ Glycosyltransferase Assay
Specifications
Content And Storage <-65°C
Quantity 50 μL