Learn More
Ultra Stomp Rocket™
SCI_ED
Catalog No. S68064
Click to view available options
Product Type:
Ultra 2-Rocket Refill Pack
the Original Stomp Rocket: Ultra 4-Rocket Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S68064 the Original Stomp Rocket: Ultra 4-Rocket Set
S68065 Ultra 2-Rocket Refill Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S68064 Supplier D & L COMPANY Supplier No. 20008
Add to Cart
Add to Cart
Students have a blast while they learn about air pressure and rocketry
Study air pressure forces and rocket technology.
  • Plastic rocket with foam tip is connected to a plastic pump
  • Step on the pump and air is forced into the rocket at high pressure causing it to soar as high as 200 ft.
  • Comes with pump, hose, adjustable-angle launching pad, and four rockets
  • Additional rockets come in packs of two
Specifications
For Use With (Application) Students have a blast while they learn about air pressure and rocketry
Includes Pump, hose, adjustable-angle launching pad, and four rockets
Material Plastic, Foam Tip
Product Type the Original Stomp Rocket: Ultra 4-Rocket Set