Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ultrafiltration Discs, 100 kDa NMW, Ultracel™ Regenerated Cellulose

Catalog No. PLHK15005 Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (Metric) Filter:
2.5 cm
4.45 cm
4.7 cm
6.35 cm
7.6 cm
9 cm
15 cm
Filtration Area:
13.4 cm2
17.3 cm2
177 cm2
28.7 cm2
4.1 cm2
41.8 cm2
63 cm2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Filter Filtration Area
PLHK15005 15 cm 177 cm2
PLHK02510 2.5 cm 4.1 cm2
PLHK06210 6.35 cm 28.7 cm2
PLHK04310 4.45 cm 13.4 cm2
PLHK04710 4.7 cm 17.3 cm2
PLHK07610 7.6 cm 41.8 cm2
PLHK09005 9 cm 63 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. PLHK15005 Supplier MilliporeSigma™ Supplier No. PLHK15005
Add to Cart
Add to Cart

100 kDa NMWL, Ultracel™ regenerated cellulose

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, Desalting
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Material (Membrane) Regenerated Cellulose
MWCO 100 kDa
Filter Code PLHK
Filtration Area 177 cm2
For Use With (Application) Protein purification; protein concentration; protein desalting
Housing Material Regenerated Cellulose (RC)
Quantity 5Pack
Type Ultrafiltration Disc
Diameter (Metric) Filter 15 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.