Learn More
Innovative Med Tech UltraFlo-R Push Button Blood Collection and Infusion Set
Description
- Sterile, single use, butterfly, multi-sample, blood collection and infusion set
- Single-handed activation retracts the needle instantly from the vein into the needle hub
- Minimizes sharp exposure
- UltraFlo needle cannula increases flow rate
- Improve patient comfort: Use 23G instead of 21G without compromising flow rate
- Approved for short-term infusion (<2 hrs)
- Unique bevel design requires lower penetration force*, providing for a more comfortable patient experience (*Compared to leading-brand standard cannula)
Specifications
Specifications
| Color | Blue |
| Material | PVC |
| Needle Gauge | 23 G |
| Sterility | Sterile |
| Latex Free | Yes |