Learn More
Foamtec UltraMOP™ IV Mop Heads
Catalog No. 19304753 Shop All Foamtec International Products
Change view
Click to view available options
Product Type:
Heavy Duty Polyester Mop Head
Polyester Mop Head
Length (English):
9 in.
14 in.
18 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (English)
19-304-753 Polyester Mop Head 9 in.
19-304-755 Polyester Mop Head 18 in.
19-304-754 Polyester Mop Head 14 in.
19-304-758 Heavy Duty Polyester Mop Head 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-304-753 Supplier Foamtec Supplier No. FS80691
Bump-proof™ wrap-around design for cleaning and disinfecting ISO Class 1 clean rooms
  • Durable snag resistant fabric construction
  • With additional abrasive mesh surface edge
  • Eliminates rollover, abrasion-related particles
  • Offers smooth, skip-free motion enhancing disinfectant laydown
  • Compatible with gamma, autoclave and ETO sterilization methods
  • Lightweight slide-on design helps for adhering mop head to Slimline™ frame
  • Available in microfiber and polyester laminated versions for stubborn contamination like hardened biofilms and disinfectant residue
Specifications
Product Type Polyester Mop Head
Length (English) 9 in.
Autoclavable Autoclavable
Includes Bump Proof Wrap around with Polyester Lamination
For Use With (Application) Cleaning and Disinfecting ISO Class 1 Cleanrooms
Material Urethane Foam