Description
- Durable snag resistant fabric construction
- With additional abrasive mesh surface edge
- Eliminates rollover, abrasion-related particles
- Offers smooth, skip-free motion enhancing disinfectant laydown
- Compatible with gamma, autoclave and ETO sterilization methods
- Lightweight slide-on design helps for adhering mop head to Slimline™ frame
- Available in microfiber and polyester laminated versions for stubborn contamination like hardened biofilms and disinfectant residue
Specifications
Specifications
| Product Type | Heavy Duty Polyester Mop Head |
| Length (English) | 18 in. |
| Autoclavable | Autoclavable |
| Includes | Bump Proof Wrap around with Heavy Duty Polyester Lamination |
| For Use With (Application) | Cleaning and Disinfecting ISO Class 1 Cleanrooms |
| Material | Urethane Foam |