Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ SSPE, 20X

Catalog No. 15591050
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-591-050 10 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-591-050 Supplier Invitrogen™ Supplier No. 15591050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UltraPure 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications.

  • Supplied in 1 L or in 10 x 1 L plastic bottles, sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2 M NaH2PO4, and 0.02 M EDTA at pH 7.4.
  • Performance and quality testing: no DNase or RNase activity detected
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Physical Form Liquid
Product Line UltraPure
Quantity 10 x 1 L

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.