Learn More
Cayman Chemical Suzetrgne 1mg
SDP
Supplier:  Cayman Chemical 407991 MG
Encompass_Preferred
A Nav1.8 inhibitor (IC50 = 0.7 nM for the human channel); selective for Nav1.8 over a panel of eight additional voltage-gated sodium channels (IC50s = >10,000 nM for all)
Catalog No. 50-294-2917