Learn More
Qsonica Ultrasonic Homogenizer Converters
Qsonica Ultrasonic Homogenizer Converter works with either the Microson XL2000 or the Sonicator 3000 Ultrasonic Homogenizers to transform electrical signals into mechanical vibrations.
$250.00 - $1183.00
Specifications
For Use With (Application) | Ideal for transform electrical signal into mechanical vibration |
---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Product Type | Price | Quantity & Availability | ||||
15-338-265
![]() |
Qsonica
K4 |
Q500 and Q700 Sonicator™ | Converter Cable |
Each for $250.00
|
|
||||
15-338-262
![]() ![]() |
Qsonica
CML4 |
Misonix™ Microson™ XL-2000 ultrasonic cell disruptor | Converter |
Each for $1,183.00
|
|
||||
Description
- For use with Microson™ Model XL2000 or Sonicator™ 3000 Ultrasonic Homgenizers
- Transforms electrical signal into mechanical vibrations
Specifications
Ideal for transform electrical signal into mechanical vibration |