Learn More
Percival Scientific Percival Scientific H15, Ultrasonic Humidifier/Dehumidifier, Precise Humidity Control, Compatible with IntellusUltra, Unit: Each
SDP
Supplier:  Percival Scientific H15
Encompass_Preferred
The PERCIVAL SCIENTIFIC H15 is an advanced ultrasonic humidifier and dehumidifier system designed for precise humidity control in scientific research environments. It actively manages relative humidity levels, ensuring stable atmospheric conditions crucial for various applications. The integrated electronic RH sensor differentiates it from competitors by providing accurate and responsive regulation, making it an essential component for Percival Scientific environmental chambers.
Catalog No. NC2309098
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.