Learn More
Marketlab Ultrasound Transducer Storage Cabinet
Catalog No. 22505051
Change view
Click to view available options
For Use With (Equipment):
Ultrasound Transducers
Catalog No. For Use With (Equipment)
22-505-051 Ultrasound Transducers
1 options
Catalog No. 22-505-051 Supplier Marketlab Supplier No. 53447
Add to Cart
Add to Cart
Ultrasound Transducer Storage Cabinet with HEPA filtration, ventilation fan, and storage for five probes.
  • 24.5 in. W x 10.75 in. D x 46.5 in. H.
  • Holds up to five probes.
  • Combi-cam lock eliminates the need for keys.
  • Includes a HEPA filter and ventilation fan with a 10 ft. cord.
  • Positive pressure fan ventilation.
  • Transparent polycarbonate window wont shatter.
  • King StarLite XL high-density polyethylene cabinet.
  • Low-profile, lightweight design.
  • Wall mountable hardware not included.
  • Clean with a damp cloth and standard hospital-grade disinfectant.
  • Recommended to replace filter annually.
Specifications
Product Type Storage Cabinet
For Use With (Equipment) Ultrasound Transducers