Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety UltraTech™ Hi-Vis Mechanics Gloves with TPR Padding

Catalog No. 19812715 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-715 Large
19-812-716 Medium
19-812-717 X-Large
19-812-718 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-812-715 Supplier MCR Safety Supplier No. MC503L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are lined with DSM Dyneema™ Diamond Technology for ANSI Level A5 cut protection. TPR padding on the back of the hand provides additional impact protection.

  • Premium goatskin leather palm is lined with DSM Dyneema™ Diamond Technology for cut protection
  • Two-way hi-vis material with TPR padding to protect the back of the hand from impacts
  • Thumb crotch is reinforced with Kevlar™ aramid fiber for additional protection in the high stress area
  • Open cuff design with pull tab for secure donning
  • ID panel on the wrist for user personalization

Specifications

ANSI Abrasion Rating ANSI 4
Back Material TPR Padding
Cuff Material Spandex
Coating Material Uncoated
Color Hi-vis Orange, Black
Cut Resistance Rating ANSI A5
Grade Premium
Hand Specific
Latex Free Yes
Product Type Mechanic's Gloves
Size Large
Finger Style Full
Glove Cuff Style Open
Liner Material DSM Dyneema™ Diamond Technology
Palm Type Goatskin Leather
Puncture Rating ANSI/ISEA 105-16 Level 3
Series MC503
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.