Learn More
MCR Safety UltraTech™ CutPro™ Hi-Vis Nitrile Foam Coated Mechanics Gloves with TPR Padding
Catalog No. 19812725 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-725 Medium
19-812-724 Large
19-812-727 X-Large
19-812-728 2X-Large
19-812-726 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-725 Supplier MCR Safety Supplier No. UT2953M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gloves feature a 13 gauge HPPE/synthetic shell with nitrile foam coating on the palm and fingers. TPR padding on the back of the hand provides additional impact protection.
  • 13-gauge HPPE/synthetic knit shell is comfortable and flexible
  • Foam nitrile coated palm and fingers enhances grip capabilities
  • TPR padding provides impact protection for the back of the hand
  • ID panel on the wrist for user personalization
  • Touch screen friendly
Specifications
ANSI Abrasion Rating ANSI 5
Back Material TPR Padding
Coating Material Nitrile Foam
Color Black, Gray, Red
Cut Resistance Rating ANSI A4
Disposable Reusable
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanic's Gloves
Size Medium
Finger Style Full
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 4
Series UT2953
Yarn Gauge 13
Show More Show Less
WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product