Learn More
MCR Safety UltraTech™ CutPro™ Nitrile Foam Coated Mechanics Gloves with TPR Padding
Catalog No. 19812722 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-722 X-Large
19-812-721 Small
19-812-723 2X-Large
19-812-720 Medium
19-812-719 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-722 Supplier MCR Safety Supplier No. UT2952XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gloves feature a 13 gauge HPPE/synthetic knit shell with a foam nitrile coating and TPR padding protection against cuts and abrasions.
  • 13-gauge HPPE/synthetic knit shell is comfortable and flexible
  • Foam nitrile coated palm and fingers enhances grip capabilities
  • TPR padding provides impact protection for the back of the hand
  • ID panel on the wrist for user personalization
  • Touch screen friendly
Specifications
ANSI Abrasion Rating ANSI 5
Back Material TPR Padding
Coating Material Nitrile Foam
Color Gray, Orange
Cut Resistance Rating ANSI A4
Disposable Reusable
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanic's Gloves
Size X-Large
Finger Style Full
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 4
Series UT2952
Yarn Gauge 13
Show More Show Less
WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product