Learn More
Epredia™ UltraVision™ Quanto Detection System HRP DAB
Catalog No. TL015QHD Shop All Epredia Products
Change view
Click to view available options
Quantity:
15 mL
60 mL
125 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL015QHD 15 mL
TL060QHD 60 mL
TL125QHD 125 mL
TL999QHD 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. TL015QHD Supplier Epredia™ Supplier No. TL015QHD
Add to Cart
Add to Cart
Epredia™ UltraVision Quanto HRP DAB delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.
UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.
  • Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
  • Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
  • Reduced cost per test — Quanto remains one of the most affordable systems available
  • Shorter incubation times — Faster run times
Includes: TL-060-QHD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Primary Antibody Amplifier Quanto(TL-060-QPB), 60mL HRP Polymer Quanto(TL-060-QPH), 60mL Ultra V Block (TA-060-UB), 60mL DAB Quanto Substrate (TA-060-QHSX), 2mL DAB Quanto Chromogen (TA-002-QHCX)
TL-125-QHD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Primary Antibody Amplifier Quanto(TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB), 125mL DAB Quanto Substrate (TA-125-QHSX), 4mL DAB Quanto Chromogen (TA-004-QHCX)
Specifications
Quantity 15 mL
Includes 15mL Hydrogen Peroxide,15mL Antibody Amplifier Quanto,15mL HRP Polymer, 15mL UltraV Block, 15mL DAB Quanto Substrate, DAB Quanto Chromogen