Learn More
AdipoGen Umbellulone
SDP
Supplier:  AdipoGen AGCN20085M010
Encompass_Preferred
Chemical. CAS 546-78-1. Formula C10H14O. MW 150.2. Isolated from Umbellularia californica Nutt. Offensive principle of the so-called "headache tree". Causes a painful cold sensation. Selective TRPA1 activator. Weak TRPM8 activator. Releases CGRP calcitonin gene-related peptide after TRPA1 activation which leads to activation of trigeminovascular pathways and CGRP-mediated neurogenic vasodilatation relevant to produce headache/migraine.
Catalog No. 50-256-2668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.