Learn More
New Pig Umbrella-Style Roof Leak Diverter Tarp Kit
SDP
Catalog No. 18389153 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
18-389-153 Yellow
1 options
This item is not returnable. View return policy
Catalog No. 18-389-153 Supplier New Pig Supplier No. TLS461YW
Add to Cart
Add to Cart
This item is not returnable. View return policy
Channel away roof or ceiling leaks before it damages inventory and machinery or causes a slip hazard.
  • Easily keep rain and snow from storming inside. Whether condensation and ice, or heavy rain from a high-wind hurricane, this solution will quickly protect your team and what you've built
  • All-in-one tarp kit hangs from a single point near the leak – just hang the hook, pull off the cover and connect the hose (included)
  • Heavy-duty vinyl-coated tarp resists punctures, UV and mildew and lasts multiple deployments
  • Pinched corners create a funnel to channel liquids to a drain or collection container without overflowing or sagging; engineered with leak-proof seams and connections
  • Brass grommets resist tearing and make installation easy; multiple hanging points add stability and preserve funnel shape
  • Extra-large drain opening and debris cage prevents clogs from crumbled ceiling tiles, drywall and roofing material
  • Choose from three colors: yellow draws attention; clear and translucent make it easy to check water flow 3/4 in. drain fitting accepts longer hoses
Specifications
Color Yellow
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov