Learn More
Apexbio Technology LLC Umeclidinium bromide 869113-09-7 50mg
Supplier: Apexbio Technology LLC A390050
Umeclidinium bromide (CAS 869113-09-7) is a potent and long-acting antagonist of muscarinic acetylcholine receptors (mAChRs) displaying high affinity for subtypes M1 M5 with Ki values of 0 16 nM 0 15 nM 0 06 nM 0 05 nM and 0 13 nM respectively It selectively targets mAChRs without observable activity at unrelated receptors or channels such as / opioid receptors sodium channels or dopamine transporters In cell-based assays using CHO cells expressing recombinant human mAChRs umeclidinium inhibits acetylcholine-induced calcium flux with pA2 values between 9 6 and 10 6 for M1 M3 receptors In murine models it reverses acetylcholine-driven bronchoconstriction Umeclidinium bromide is broadly applied in research focused on pulmonary disease mechanisms and drug development
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.