Learn More
TARGETMOL CHEMICALS INC UMIBECESTAT 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T13253
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Umibecestat is an inhibitor of beta-site amyloid precursor protein cleaving enzyme-1 (BACE-1) with IC50s of 11 nM and 10 nM for human BACE-1 and mouse BACE-1 respectively. Umibecestat can be used for the research of alzheimer's disease. purity: 100%
Catalog No. 50-258-1364
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.