Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

All-Fi Standard UN Rated All-Fiber Drum
SDP

Catalog No. 18366525 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (English):
12 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-366-525 12 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-525 Supplier All-Fi Standard Supplier No. DRM798
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Completely incinerable 12-gallon fiber drum for storing or disposing of dry materials.

  • Storage Capacity 12 gal.
  • UN1G/X60/S
  • Interior dimensions: 12.5 in. diameter and 22.5 in. height
  • All-fiber, UN Rated drum is ideal for storing or disposing of dry materials.
  • Flat top and bottom make stacking simple and safe
  • Telescoping cover slips over top, so you can utilize entire inside capacity
  • No need to empty drum to burn contents for waste reduction or fuels blending, as drum is completely incinerable
  • Ideal for pelletized resins, colorants and other high-volume dry materials

Specifications

Product Type Fiber Drum
Capacity (English) 12 gal.
Capacity (Metric) 12 gal.
Color Brown
Material Linerboard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.