Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UNC 0638
GREENER_CHOICE

Catalog No. 434310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-431-0 10 mg
43-435-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-431-0 Supplier Tocris Bioscience™ Supplier No. 4343/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective G9a and GLP inhibitor

Selective inhibitor of G9a and GLP histone lysine methyltransferases (IC50 values are < 15 nM and 19 nM for G9a and GLP respectively, and > 10000 nM for a range of other histone methyltransferases). Potently inhibits dimethylation of H3K9 in MCF-7 cells (IC50 = 70 nM). Cell permeable.

Chemical Identifiers

CAS 1255517-77-1
Molecular Formula C30H49N5O3

Specifications

Quantity 10 mg
Formula Weight 509.73
Purity >99%
Molecular Weight (g/mol) 527.754
Chemical Name or Material UNC 0638
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.