Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UNC 2881
GREENER_CHOICE

Catalog No. p-7161687
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-372-5 25 mg
65-375 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-372-5 Supplier Tocris Bioscience™ Supplier No. 6537/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent Mer kinase inhibitor

UNC 2881 is a potent Mer kinase inhibitor. Displays >55-fold selectivity for Mer kinase over other TAM receptor tyrosine kinase family members (IC50 values are 4.3 nM, 0.25 μM and 0.36 μM for Mer, Tyro3 and Alk, respectively). Inhibits Mer phosphorylation in vitro. Also inhibits platelet aggregation in response to Type I collagen.

Specifications

Chemical Name or Material 2-(Butylamino)-4-[(trans-4-hydroxycyclohexyl)amino]-N-[[4-(1H-imidazol-1-yl)phenyl]methyl]-5-pyrimidinecarboxamide
CAS 1493764-08-1
Quantity 25 mg
Molecular Formula C25H33N7O2
Formula Weight 463.58
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.