Learn More
TARGETMOL CHEMICALS INC UNC0638 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3257
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UNC0638 is an inhibitor of (beta)-protein lysine methyltransferases G9a(IC50<15 nM) and GLP(IC50=19 nM) with excellent potency and selectivity over a wide range of epigenetic and non-epigenetic targets. purity: 99%
Catalog No. 50-258-0328
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.