Learn More
Apexbio Technology LLC UNC2250, 5mg. Cas: 1493694-70-4 MFCD: MFCD27992060. Mer inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B20605
Encompass_Preferred
UNC2250 is a highly selective inhibitor of Mer receptor tyrosine kinase with IC50 value of 1.7nM [1].UNC2250 shows a potent inhibitory activity against Mer with IC50 value of 1.7nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5301
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.