Learn More
Selleck Chemical LLC UNC3866 5mg 1872382-47-2
SDP
Supplier:  Selleck Chemical LLC S83595MG
Encompass_Preferred
UNC3866 is a potent antagonist of the methyllysine (Kme) reading function of the Polycomb CBX and CDY families of chromodomains. UNC3866 binds the chromodomains of CBX4 and CBX7 most potently, with a K(d) of ?100 nM for each, and is 6- to 18-fold selective as compared to seven other CBX and CDY chromodomains. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5982
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.