Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112960
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112960 Supplier Invitrogen™ Supplier No. PA5112960
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC50 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

UNC50 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC50
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene UNC50
Gene Accession No. Q53HI1
Gene Alias 1110002A21Rik; geal-6 membrane-associated high-copy suppressor 1; GMH1; hGMH1; HSD23; HSD-23; hypothetical protein LOC513629; PDLs22; Periodontal ligament-specific protein 22; protein GMH1 homolog; Protein unc-50 homolog; UNC50; unc-50 homolog; unc-50 homolog (C. elegans); unc-50 inner nuclear membrane RNA binding protein; unc-50 related; unc-50 related protein (UNCL); unc-50-like protein; UNCL; uncoordinated-like protein; URP; zgc:56114
Gene Symbols UNC50
Host Species Rabbit
Immunogen Recombinant Human Protein unc-50 homolog protein (15-82AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25972
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.