Learn More
Selleck Chemical LLC UNC6852 5mg
SDP
Supplier:  Selleck Chemical LLC S96595MG
Encompass_Preferred
UNC6852 is a selective degrader that targets polycomb repressive complex 2 (PRC2) with IC50 of 247 nM for EED. UNC6852 is based on PROTAC and contains an EED226-derived ligand and a ligand for VHL. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.