Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanedinitrile 95.0+%, TCI America™
SDP

Catalog No. U005925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U005925ML 25 mL
U00595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U005925ML Supplier TCI America Supplier No. U005925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71172-36-6
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.279
MDL Number MFCD00039506
InChI Key ISIQQQYKUPBYSL-UHFFFAOYSA-N
Synonym 1,9-Dicyanononane
PubChem CID 247906
IUPAC Name undecanedinitrile
SMILES C(CCCCC#N)CCCCC#N

Specifications

Color Colorless
Boiling Point 190°C
Quantity 25 mL
UN Number 3276
Formula Weight 178.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecanedinitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.