Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanophenone 98.0+%, TCI America™
SDP

Catalog No. U005325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U005325G 25 g
U0053500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U005325G Supplier TCI America Supplier No. U005325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4433-30-1
Molecular Formula C17H26O
Molecular Weight (g/mol) 246.394
MDL Number MFCD00051548
InChI Key LHJBFOGCFZHBAJ-UHFFFAOYSA-N
Synonym undecanophenone, n-undecanophenone, 1-phenylundecanone, 1-undecanone, 1-phenyl, decyl phenyl ketone, 1-phenyl-1-undecanone, 1-phenyl-undecan-1-one, 1-phenyl-1-undecanone #, acmc-209jx6
PubChem CID 78167
IUPAC Name 1-phenylundecan-1-one
SMILES CCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Undecanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.