Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Under BSC Mount Serological Pipette Holder
SDP

Catalog No. 01259154
Change view
Click to view available options
Positions:
4 Pockets
5 Pockets
6 Pockets
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
01-259-154 4 Pockets
01-259-193 5 Pockets
01-259-178 6 Pockets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-154 Supplier Poltex Supplier No. SRPIPUNDR4M
Add to Cart
Add to Cart
N/A

Designed for mounting under a tissue culture hood or BSC in horizontal position. Available in 4, 5, or 6 pocket capacity with Magnet Mount and durable PETG construction.

  • Specifically designed for mounting under a tissue culture hood or BSC in a horizontal position.
  • Choose from 4, 5, or 6 pocket capacities.
  • Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Serological Pipets, Pipets, Glass and Plastic Pipets
Material PETG
Positions 4 Pockets
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Application) Undermount Serological Pipette Holder
Mounting Magnet Mount
Dimensions (L x W x H) 16.3 x 11.85 x 3.5 in.
Weight (English) 4 lb.
Includes Magnets for Mounting
Product Type Serological Pipet Holder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.