Learn More
Thomas Scientific Absorbent Bench Underpad, Bench pads, 16.5 x 23 in./42 x 58cm., 50 per pack
SDP
Supplier:  Thomas Scientific 1158J48
Encompass_Preferred
Absorbs up to 500ml per pad. Filled with 100% fluffed cellulose. Blue polyethylene liner.
Catalog No. NC1588452