Learn More
Med Vet International VACUETTE Venous Blood Collection Tube K2 EDTA Additive 4 mL Pull Cap Polyethylene Terephthalate (PET) Tube, 454209, 1200/Case
Supplier: Med Vet International 505509CS
The interior of these Vacuette tubes is coated with K2 EDTA – an anticoagulant that prevents blood from clotting by inhibiting the action of clotting factors Pull caps are lavender with black rings Dimensions:13 x 75 millimeters Volume: 3 mL Clear, silica-coated tube is polyethylene terephthalate (PET)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.