Learn More
Thermo Scientific™ Unifilter Direct-Connection Filter System Components
Catalog No. 27000
Change view
Click to view available options
Diameter (Metric) Outer:
3.0/2.1 mm
4.0/4.6 mm
Type:
Unifilter Direct Connection Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Type
27000 4.0/4.6 mm Unifilter Direct Connection Holder
27002 3.0/2.1 mm Unifilter Direct Connection Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27000 Supplier Thermo Scientific™ Supplier No. 27000
Add to Cart
Add to Cart
Minimize downtime with these easily replaceable filter system components.
Minimize downtime. Thermo Scientific™ Unifilter Direct-Connection Filter System Components are easily replaced.
  • Replaceable 0.5μm drop-in filter enhances column lifetime and improved performance
  • Holder attached directly to the inlet of analytical system for maximum convenience
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 4/4.6 mm
For Use With (Equipment) Unifilter Direct-Connection HPLC Filter Systems
Diameter (Metric) Outer 4.0/4.6 mm
Description Unifilter Direct-Connection Filter Holder; 4.0/4.6 mm
Type Unifilter Direct Connection Holder
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.