Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protective Industrial Products Uniform Technology™ Altessa Grid Navy Frock

Catalog No. 17010679 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-679 2X-Large
17-010-671 Small
17-010-672 Medium
17-010-673 Large
17-010-670 X-Large
17-010-678 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-010-679 Supplier Protective Industrial Products Supplier No. CFRZC74NV2XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reusable cleanroom frock features raglan sleeves and ESD knit cuffs for use in ISO 5 or higher compatible environments.

  • Navy
  • Fabric 99% Polyester 1% Carbon
  • ISO 5 (Class 100)
  • Features raglan sleeve, military collar, coil zipper at front opening, snaps at collar and snap 2 in. below bottom of zipper, sleeves with ESD knit cuff

Specifications

Size 2X-Large
Material Polyester, Carbon
Color Navy
Disposable Reusable
Sterility Non-sterile
Closure Type Zipper with Snaps
Cuff Style ESD Knit
Collar Type Military
Latex Free Yes
Autoclavable Autoclavable
Cleanroom Class Class 100 (ISO 5)
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Electronics and Aerospace, Food Processing, Controlled Environments, Film Coating and Imaging Manufacturing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.