Learn More
Protective Industrial Products Uniform Technology™ Altessa Grid Navy Hood
Catalog No. 17010640 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-640 Large
17-010-638 Small
17-010-639 Medium
17-010-637 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-010-640 Supplier Protective Industrial Products Supplier No. CHM74NVL
Add to Cart
Add to Cart
Reusable Cleanroom Hood features open face, snaps under chin and each side at temple, vertical and horizontal snaps in back, and rolled hem.
  • Navy
  • Fabric 99% Polyester 1% Carbon
  • ISO 5 (Class 100)
  • Features open face, snaps under chin and each side at temple, vertical and horizontal snaps in back, roll hem
Specifications
Product Type Hood
Size Large
Material Polyester, Carbon
Color Navy
Closure Type Snap
Disposable Reusable
Latex Free Yes
Sterility Non-sterile
Cleanroom Class Class 100 (ISO 5)