Learn More
Protective Industrial Products Uniform Technology™ Altessa Grid White Coverall
Catalog No. 17010680 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-680 X-Small
17-010-682 Small
17-010-683 Medium
17-010-684 Large
17-010-681 X-Large
17-010-690 2X-Large
17-010-689 3X-Large
17-010-688 4X-Large
17-010-687 5X-Large
17-010-686 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17-010-680 Supplier Protective Industrial Products Supplier No. CC124574WHXS
Only null left
Add to Cart
Add to Cart
Reusable Cleanroom Coverall features raglan sleeves, military collar, color coded hang loop, coil zipper, snaps at top of zipper, ESD knit cuffs at sleeve opening, and snaps at leg hems and backs of legs.
  • Navy
  • Fabric 99% Polyester 1% Carbon
  • ISO 5 (Class 100)
  • Features raglan sleeves, military collar, color coded hang loop, coil zipper, snaps at top of zipper, ESD knit cuffs at sleeve opening, snaps at leg hems and backs of legs
Specifications
Product Type Cleanroom Coveralls
Size X-Small
Gender Unisex
Material Polyester, Carbon
Color White
Disposable Reusable
Ankle Style Snap
Cuff Style Knit
Neck Style Collared
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100 (ISO 5)