Learn More
Protective Industrial Products Uniform Technology™ Sitewear Top ESD Microdenier
Catalog No. 17456164 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Catalog No. Size
17-456-164 Small
1 options
Catalog No. 17-456-164 Supplier Protective Industrial Products Supplier No. HSCTM348NVS
Add to Cart
Add to Cart
Reusable Sitewear Top with long raglan sleeve, placket front with interfacing, knit cuffs, bound neck, and slits at bottom of side seams.
  • Reusable Sitewear Top long raglan sleeve, placket front with interfacing, knit cuffs, bound neck, slits at bottom of side seams.
  • ESD Microdenier 99% Polyester 1% Carbon
Specifications
Size Small
Gender Unisex
Material Polyester, Carbon
Color Navy
Neck Style Gripper
Seam Type Serged
Sleeve Style Raglan Sleeve
Disposable Reusable
Latex Free Yes
No. of Pockets None
Product Type Long Sleeve Shirt
Sterility Non-sterile