Learn More
MilliporeSigma™ Supelco™ Union bore 0.010 in., thread 10-32
Catalog No. 111011616 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Carbon-filled PEEK
Catalog No. Material
11-101-1616 Carbon-filled PEEK
1 options
Catalog No. 11-101-1616 Supplier MilliporeSigma™ Supelco™ Supplier No. 57658
Add to Cart
Add to Cart
The PEEK zero dead volume unions are to be operated at 5000 psi (352 kg/cm2). They include Fingertight III nuts and ferrules.
TRUSTED_SUSTAINABILITY
Specifications
Material Carbon-filled PEEK