Learn More
MilliporeSigma™ Supelco™ Union, Reducing for connecting 1/16 to 1/32 in., Bore Size 0.25 mm
Catalog No. 111011576 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-101-1576 1/Pk.
1 options
Catalog No. 11-101-1576 Supplier MilliporeSigma™ Supelco™ Supplier No. 59025U
Add to Cart
Add to Cart
Stainless steel, unless noted otherwise. Nuts and ferrules included.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Material Stainless Steel
Valco is a registered trademark of Valco Instruments Co. Inc.