Learn More
United Scientific™ UniSafe™ Assorted Vented Wash Bottles, LDPE, 500 mL

Shop All United Scientific Supplies, Inc Products


Description
Based on our best sellers, we have added an assorted pack to our labForce™ vented 500 mL wash bottle line. Assorted pack of six includes: Acetone, Ethanol, Isopropanol, Distilled Water, Sodium Hypochlorite, and Write-In (create your own label).
labForce™ vented wash bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.They are pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200. Intended for use only with LDPE and PP compatible solvents.
- Assorted pack: Acetone, Ethanol, Isopropanol, Distilled Water, Sodium Hypochlorite, and Write-In
- Industrial quality low density polyethylene (LDPE) construction with long lasting strength, GHS compliant
- Preprinted bottles include: chemical name and formula, NFPA diamond, health hazards, and CAS number
- Polypropylene stem closure and draw tube
- Safety vented and color coded for quick solvent recognition
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Material | LDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Chemical Label | Assorted |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 16 oz. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.