Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniSafe™ Vented Wash Bottle, 1000 mL
SDPSCI_ED

Catalog No. S80005 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
S80005 Sodium Hypochlorite
S80006 Isopropanol
S80007 Distilled Water
S80008 Methanol
S80009 Ethanol
S80010 Acetone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S80005 Supplier United Scientific™ Supplier No. UN370079
Add to Cart
Add to Cart

UniSafe Vented Wash Bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.

  • United Scientific™ UniSafe™ vented wash bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.
  • They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
  • Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
  • Intended for use only with LDPE and PP compatible solvents.
  • Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
  • Polypropylene stem closure and draw tube
  • Safety vented and color coded for quick solvent recognition
  • 4-color printing, 1000 mL capacity
  • Available in 6 different varieties.
  • Sold as packs of 4 bottles.

Specifications

Capacity (Metric) 1000 mL
Capacity (English) 33.8 oz.
Color Translucent
Material LDPE
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Safety-vented Wash Bottle
Chemical Label Sodium Hypochlorite
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.