Learn More
United Scientific™ UniSafe™ Vented Wash Bottle, 1000 mL

Shop All United Scientific Supplies, Inc Products


Description
- United Scientific™ UniSafe™ vented wash bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.
- They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
- Intended for use only with LDPE and PP compatible solvents.
- Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
- Polypropylene stem closure and draw tube
- Safety vented and color coded for quick solvent recognition
- 4-color printing, 1000 mL capacity
- Available in 6 different varieties.
- Sold as packs of 4 bottles.
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Capacity (English) | 33.8 oz. |
| Color | Translucent |
| Material | LDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Mouth | Wide, Threaded |
| Shape | Cylindrical |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.