Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniSafe™ Vented Wash Bottle
SDPSCI_ED

Catalog No. S00316 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Chemical Label:
Acetone
Acetonitrile
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite
Toluene
Water
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
S00316 Water
S00317 Toluene
S00318 Sodium Hypochlorite
S00319 Methyl Ethyl Ketone
S00320 Methanol
S00321 Isopropanol
S00322 Ethyl Acetate
S00323 Ethanol
S00324 Distilled Water
S00325 Dichloromethane
S00326 Acetonitrile
S00327 Acetone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. S00316 Supplier United Scientific™ Supplier No. UN370061
Add to Cart
Add to Cart
This item is not returnable. View return policy

UniSafe™ vented wash bottles made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.

Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200. Intended for use only with LDPE and PP compatible solvents.

  • Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
  • Preprinted bottles include: chemical name and formula, NFPA diamond, health hazards, and CAS number
  • Polypropylene stem closure and draw tube
  • Safety vented and color coded for quick solvent recognition
  • 4-color printing, 500 mL (16 oz.) capacity

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Blue
For Use With (Application) LDPE and PP-compatible Solvents
Material LDPE
Packaging 6/Pk.
Sterility Non-sterile
Certifications/Compliance GHS and OSHA
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Threaded
Shape Cylindrical
Product Type Vented Wash Bottle
Chemical Label Water
Closure Material Polypropylene
Closure Type Vented Screw Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.