Learn More
Description
Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200. Intended for use only with LDPE and PP compatible solvents.
- Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
- Preprinted bottles include: chemical name and formula, NFPA diamond, health hazards, and CAS number
- Polypropylene stem closure and draw tube
- Safety vented and color coded for quick solvent recognition
- 4-color printing, 500 mL (16 oz.) capacity
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Green |
| For Use With (Application) | LDPE and PP-compatible Solvents |
| Material | LDPE |
| Packaging | 6/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Threaded |
| Chemical Label | Methanol |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

