Learn More
United Scientific™ UniSafe™ Vented Wash Bottle
SDPSCI_ED
Catalog No. S80011 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Chemical Label:
Custom Write-on
Deionized Water
Soap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
S80011 Custom Write-on
S80012 Deionized Water
S80013 Soap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S80011 Supplier United Scientific™ Supplier No. UN370070
Only null left
Add to Cart
Add to Cart
UniSafe Vented Wash Bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.
  • United Scientific™ UniSafe™ vented wash bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.
  • They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
  • Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
  • Intended for use only with LDPE and PP compatible solvents.
  • Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
  • Polypropylene stem closure and draw tube
  • Safety vented and color coded for quick solvent recognition
  • 4-color printing, 500 mL capacity
  • Available in 15 different varieties.
  • Sold as packs of 6 bottles.
Specifications
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Translucent
Material LDPE
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide, Threaded
Shape Cylindrical
Product Type Safety-vented Wash Bottle
Chemical Label Custom Write-on
Autoclavable Not Autoclavable