Learn More
Simport™ Scientific UNISETTE™ Biopsy Processing/Embedding Cassette
Catalog No. 22045022 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-045-022 White
22-045-023 Pink
22-045-024 Green
22-045-025 Yellow
22-045-026 Blue
22-045-027 Peach
22-045-028 Tan
22-045-029 Gray
22-045-030 Lilac
22-045-031 Orange
22-045-032 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 missing translation for 'options'
Catalog No. 22-045-022 missing translation for 'mfr' Simport™ Scientific missing translation for 'supplierNo' M5062
Add to Cart
Add to Cart
Secure specimens during processing with UNISETTE™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Writing Face Angle 35°
Color White
Material Acetal
Product Type Biopsy Processing/Embedding Cassette
For in vitro diagnostic (IVD) use only.